accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOHOHHOHOOOH3COOCH3

CAS RN 77307-50-7

IUPAC Name: 4-(acetyloxy)-6-{[5,7-dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-3-yl]oxy}-5-hydroxy-2-methyloxan-3-yl acetate
Synonyms: 4-(acetyloxy)-6-{[5,7-dihydroxy-2-(4-hydroxyphenyl)-4-oxochromen-3-yl]oxy}-5-hydroxy-2-methyloxan-3-yl acetate
Molecular Weight (g/mol): 516.46
Molecular Formula: C25H24O12
InChi Key: SXOZSDJHGMAEGZ-UHFFFAOYNA-N
SMILES: CC1OC(OC2=C(OC3=CC(O)=CC(O)=C3C2=O)C2=CC=C(O)C=C2)C(O)C(OC(C)=O)C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results