Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SL 0101-1
GREENER_CHOICESDP

Catalog No. 22501 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-501 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-501 Supplier R&D Systems™ Supplier No. 2250/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective ribosomal S6 kinase (RSK) inhibitor

Selective inhibitor of ribosomal S6 kinase (RSK) (IC50 = 89 nM for RSK2). Does not inhibit upstream kinases such as MEK, Raf and PKC. Inhibits the growth of MCF-7 human breast cancer cells with no effect on the normal breast cell line.

Chemical Identifiers

CAS 77307-50-7
Molecular Formula C25H24O12

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 516.46
Purity >98%
Molecular Weight (g/mol) 516.455
Chemical Name or Material SL 0101-1
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.