accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnOSOOO

CAS RN 7733-02-0

IUPAC Name: zinc(2+) sulfate
Synonyms: zinc sulfate zinc(II) sulfate ZnSO4 zinc sulfate anhydrous
Molecular Weight (g/mol): 161.44
Molecular Formula: O4SZn
InChi Key: NWONKYPBYAMBJT-UHFFFAOYSA-L
SMILES: [Zn++].[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (5)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)