Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc sulfate concentrate, For 1L standard solution, 0.1 M ZnSO4, Solstice

Catalog No. 60046058 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-058 6 x 1 Ea.
60-017-86 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-058 Supplier Solstice Supplier No. 320476X1EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for 1L standard solution, 0.1 M ZnSO4

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7733-02-0 , 7732-18-5
Molecular Formula O4SZn
Molecular Weight (g/mol) 161.44
MDL Number MFCD00011302
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
Synonym zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
PubChem CID 24424
ChEBI CHEBI:35176
IUPAC Name zinc;sulfate
SMILES [Zn++].[O-]S([O-])(=O)=O

Specifications

Physical Form Concentrate
CAS Max % 35.943%, 64.0570%
Quantity 6 x 1 Ea.
Packaging Ampoule
Linear Formula ZnSO4
UN Number UN3082
Formula Weight 161.45
Chemical Name or Material Zinc sulfate concentrate, For 1L standard solution, 0.1 M ZnSO4
Hazard Statement P273-P280-P305 + P351 + P338-P501
Precautionary Statement H318-H410
ShelfLife 1260 days from date of manufacture
DOTInformation 9 / PGIII
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.