Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate Solution, 0.05M, Solstice

Catalog No. 6002509 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-09 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-025-09 Supplier Solstice Supplier No. 344481L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 0.05 M ZnSO4

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7733-02-0
Molecular Formula O4SZn
Molecular Weight (g/mol) 161.44
MDL Number MFCD00011302
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
Synonym zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
PubChem CID 24424
ChEBI CHEBI:35176
IUPAC Name zinc;sulfate
SMILES [Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 L
Linear Formula ZnSO4
UN Number NONH for all modes of transport
Formula Weight 161.45g/mol
Chemical Name or Material Zinc sulfate solution
Hazard Statement P273-P305 + P351 + P338
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.