accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77341-67-4

IUPAC Name: 4-{[5-methyl-2-(propan-2-yl)cyclohexyl]oxy}-4-oxobutanoic acid
Synonyms: menthyl succinate
Molecular Weight (g/mol): 256.34
Molecular Formula: C14H24O4
InChi Key: BLILOGGUTRWFNI-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OC(=O)CCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results