CAS RN 7758-02-3
Potassium bromide, 99+%, for spectroscopy, IR grade
CAS: 7758-02-3 Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M Synonym: potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095 PubChem CID: 253877 ChEBI: CHEBI:32030 SMILES: [K+].[Br-]
Potassium Bromide, Crystal, ACS, 99%, Spectrum™ Chemical
CAS: 7758-02-3 Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M IUPAC Name: potassium bromide SMILES: [K+].[Br-]
Potassium Bromide, Crystal, IR Grade, Spectrum™ Chemical
CAS: 7758-02-3 Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M IUPAC Name: potassium bromide SMILES: [K+].[Br-]
Potassium Bromide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7758-02-3 Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M Synonym: potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095 PubChem CID: 253877 ChEBI: CHEBI:32030 IUPAC Name: potassium bromide SMILES: [K+].[Br-]
Potassium Bromide, Granular, Purified, Approx. 98%, Spectrum™ Chemical
CAS: 7758-02-3
Potassium Bromide, ACS Reagent Grade, Ricca Chemical
CAS: 7758-02-3 Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M Synonym: potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095 PubChem CID: 253877 ChEBI: CHEBI:32030 IUPAC Name: potassium bromide SMILES: [K+].[Br-]
Potassium Bromide, Certified, 10.0% (w/v) ±0.5% (w/v), LabChem™
CAS: 7758-02-3 Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M Synonym: potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095 PubChem CID: 253877 ChEBI: CHEBI:32030 IUPAC Name: potassium bromide SMILES: [K+].[Br-]
Potassium Bromide Solution, For Mercury, LabChem™
CAS: 7758-02-3 Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M Synonym: potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095 PubChem CID: 253877 ChEBI: CHEBI:32030 IUPAC Name: potassium bromide SMILES: [K+].[Br-]
Potassium Bromide, puriss., Solstice
Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M SMILES: [K+].[Br-]
Bromide bromate Standard Solution, 0.25M, Solstice
CAS: 7758-02-3 Molecular Formula: BrK Molecular Weight (g/mol): 119.00 MDL Number: MFCD00011358 InChI Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M IUPAC Name: potassium bromide SMILES: [K+].[Br-]