Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003721 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-721 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-721 Supplier Avantor Supplier No. 299701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7758-02-3 , 7732-18-5
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 118.99
Percent Purity 99.4%
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.