Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, Crystal, IR Grade, Spectrum™ Chemical
SDP

Catalog No. 18605475 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
6 x 25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-475 100 g Amber Glass Bottle
18-605-473 25 g Amber Glass Bottle
18-605-474 6 x 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-605-475 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1225100GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1225, 7758-02-3, KBr

Spectrum™ Chemical Potassium Bromide, Crystal, IR Grade is used widely as infrared optical windows and components for general spectroscopy because it is transparent from the near ultraviolet to long-wave infrared wavelengths and has no significant optical absorption lines in its high transmission region.

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

pH 5.0 to 8.8
CAS Max % 1
Physical Form Crystals or Needles
Quantity 100 g
Grade IR
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Bromide, Crystal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.