accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OO

CAS RN 7774-44-9

IUPAC Name: cyclohexyl 3-methylbutanoate
Synonyms: cyclohexyl isovalerate
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: SQPOKBBCNZIWFI-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)OC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results