accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOOOOSOOO

CAS RN 7775-27-1

IUPAC Name: disodium [(sulfonatoperoxy)sulfonyl]oxidanide
Synonyms: sodium persulfate
Molecular Weight (g/mol): 238.09
Molecular Formula: Na2O8S2
InChi Key: CHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (17)
  • (5)
Quantity 
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (7)
Grade 
  • (2)
  • (4)
  • (1)
  • (7)
  • (5)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (1)
  • (7)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (3)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (17)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (7)
  • (2)
  • (4)
  • (1)
  • (7)
  • (5)
  • (3)
  • (3)
  • (3)
  • (1)
  • (7)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (4)