Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Peroxidisulfate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1066090500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1066090500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1066090500 Supplier MilliporeSigma Supplier No. 1.06609.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity EMSURE™ grade sodium peroxidisulfate for analytical laboratory applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
pH 3.5 to 3.8 (100g/L, H2O, 20°C)
Physical Form Solid
Quantity 500 g
Solubility Information 545g/L
Formula Weight 238.11
Percent Purity >99.0%
Grade Not Available
Packaging Plastic Bottle
Density 1.2 g/3 (20 °C)
Chemical Name or Material Sodium Persulfate
Show More Show Less
EINECSNumber 231-892-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.