Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Persulfate, BAKER™, J.T. Baker™

Catalog No. 02004184 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-184 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-184 Supplier Avantor Supplier No. V03507
Add to Cart
Add to Cart

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 500 g
Formula Weight 238.03
Packaging Poly Bottle
Specific Gravity 1L = 2.40kg
Chemical Name or Material Sodium Persulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.