accessibility menu, dialog, popup

UserName

Viewing profile as user:
IFFFFFFFFFFFFFFFFFFFFSOOO

CAS RN 77758-89-5

IUPAC Name: (1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)(phenyl)iodanium trifluoromethanesulfonate
Synonyms: (1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)(phenyl)iodanium triflate
Molecular Weight (g/mol): 772.13
Molecular Formula: C15H5F20IO3S
InChi Key: LQSJCHNACSIGQK-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)[I+]C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results