accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7778-54-3

IUPAC Name: calcium dihypochlorite
Synonyms: calcium hypochlorite
Molecular Weight (g/mol): 142.98
Molecular Formula: CaCl2O2
InChi Key: ZKQDCIXGCQPQNV-UHFFFAOYSA-N
SMILES: [Ca++].[O-]Cl.[O-]Cl
Molecular Weight (g/mol) 
  • (4)
  • (5)
Physical Form 
  • (2)
  • (3)
  • (3)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (6)
  • (1)
  • (4)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)