Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hypochlorite, Tablets, 70% Available Chlorine (Tech.), Thermo Scientific™

Catalog No. AC199030010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC199030010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC199030010 Supplier Thermo Scientific Chemicals Supplier No. 199030010

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
Molecular Weight (g/mol) 142.976
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Synonym calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor
PubChem CID 24504
IUPAC Name calcium;dihypochlorite
SMILES [O-]Cl.[O-]Cl.[Ca+2]

Specifications

Color White
pH ∼11.8 (1% soln.)
Physical Form Tablets
Quantity 1 kg
Linear Formula Ca(OCl)2
Fieser 11,107
Merck Index 15, 1676
Formula Weight 142.98
Percent Purity ≥70% (available Cl)
Grade Technical
Density 2.35
Chemical Name or Material Calcium hypochlorite, tech.
Show More Show Less
Hazard Category Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.