accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH3CH2

CAS RN 7778-96-3

IUPAC Name: 3,7-dimethyloct-7-en-1-yl 3-methylbutanoate
Synonyms: rhodinyl isovalerate
Molecular Weight (g/mol): 240.39
Molecular Formula: C15H28O2
InChi Key: OZAWINZSOFVOBJ-UHFFFAOYNA-N
SMILES: CC(C)CC(=O)OCCC(C)CCCC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results