CAS RN 7784-19-2
IUPAC Name:
triammonium hexafluoroalumanetriuide
Synonyms:
triammonium hexafluoroalumanetriuide
Molecular Weight (g/mol):
195.09
Molecular Formula:
AlF6H12N3
InChi Key:
OYHBNKHFKHBTRQ-UHFFFAOYSA-K
SMILES:
[NH4+].[NH4+].[NH4+].F[Al-3](F)(F)(F)(F)F
1
–
1
of
1
results
1
Ammonium hexafluoroaluminate, 98%
CAS: 7784-19-2 Molecular Formula: AlF6H12N3 Molecular Weight (g/mol): 195.09 MDL Number: MFCD00042144 InChI Key: OYHBNKHFKHBTRQ-UHFFFAOYSA-K Synonym: ammonium hexafluoroaluminate,ammonium cryolite,ammonium fluoaluminate,ammonium aluminum fluoride,triammonium hexafluoroaluminate,triammonium aluminum hexafluoride,triammoniumhexafluoroaluminate 3-,hexafluoroaluminate 3-triammonium,aluminum ammonium fluoride al nh4 3f6,ammonium hexafluoroaluminate nh4 3alf6 PubChem CID: 24565 IUPAC Name: triazanium;hexafluoroaluminum(3-) SMILES: [NH4+].[NH4+].[NH4+].F[Al-3](F)(F)(F)(F)F