CAS RN 7789-02-8
Chromium(III) nitrate nonahydrate, 99%
CAS: 7789-02-8 MDL Number: MFCD00149659 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N Synonym: chromium iii nitrate nonahydrate,unii-d2806iol1l,nitric acid, chromium 3+ salt, nonahydrate,acmc-20alg5,bis 7-chlorophenyl sulfone,cr.3no3.9h2o,ksc495a9j,chromium trinitrate, nonahydrate,chromium iii nitrate, nonahydrate,chromium 3+ nonahydrate trinitronate PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Chromium(III) nitrate nonahydrate, 98.5%
CAS: 7789-02-8 Molecular Formula: CrN3O9 Molecular Weight (g/mol): 238.01 MDL Number: MFCD00149659 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N Synonym: chromium iii nitrate nonahydrate,unii-d2806iol1l,nitric acid, chromium 3+ salt, nonahydrate,acmc-20alg5,bis 7-chlorophenyl sulfone,cr.3no3.9h2o,ksc495a9j,chromium trinitrate, nonahydrate,chromium iii nitrate, nonahydrate,chromium 3+ nonahydrate trinitronate PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Chromium Nitrate, Technical, Spectrum™ Chemical
CAS: 7789-02-8 Molecular Weight (g/mol): 400.15
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL
CAS: 7789-02-8 Molecular Formula: CrN3O9 Molecular Weight (g/mol): 238.01 MDL Number: MFCD00010944 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Chromium(III) nitrate nonahydrate, 99.99% min (metals basis)
CAS: 7789-02-8 Molecular Formula: CrN3O9 Molecular Weight (g/mol): 238.01 MDL Number: MFCD00149659 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N Synonym: chromium iii nitrate nonahydrate,unii-d2806iol1l,nitric acid, chromium 3+ salt, nonahydrate,acmc-20alg5,bis 7-chlorophenyl sulfone,cr.3no3.9h2o,ksc495a9j,chromium trinitrate, nonahydrate,chromium iii nitrate, nonahydrate,chromium 3+ nonahydrate trinitronate PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Chromium nitrate nonahydrate, 99.99%, (trace metal basis)
CAS: 7789-02-8 Molecular Formula: CrN3O9·9H2O Molecular Weight (g/mol): 400.14 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N Synonym: chromium iii nitrate nonahydrate,unii-d2806iol1l,nitric acid, chromium 3+ salt, nonahydrate,acmc-20alg5,bis 7-chlorophenyl sulfone,cr.3no3.9h2o,ksc495a9j,chromium trinitrate, nonahydrate,chromium iii nitrate, nonahydrate,chromium 3+ nonahydrate trinitronate PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Chromium ICP Standard, 1000 ppm Cr in 3% HNO3, Ricca Chemical
CAS: 7789-02-8 Molecular Formula: CrN3O9 Molecular Weight (g/mol): 238.01 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Chromium ICP-MS Standard, 1000 ppm Cr in 3% HNO3, Ricca Chemical
CAS: 7789-02-8 Molecular Formula: CrN3O9 Molecular Weight (g/mol): 238.01 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Chromium(III) Nitrate Nonahydrate, puriss. p.a., ≥98.0%, Solstice
CAS: 7789-02-8 Molecular Formula: CrN3O9 Molecular Weight (g/mol): 238.01 MDL Number: MFCD00149659 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N Synonym: chromium iii nitrate nonahydrate,unii-d2806iol1l,nitric acid, chromium 3+ salt, nonahydrate,acmc-20alg5,bis 7-chlorophenyl sulfone,cr.3no3.9h2o,ksc495a9j,chromium trinitrate, nonahydrate,chromium iii nitrate, nonahydrate,chromium 3+ nonahydrate trinitronate PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Fisher Science Education™ Chromium Nitrate, Nonahydrate
CAS: 7789-02-8 Molecular Formula: CrN3O9 Molecular Weight (g/mol): 238.01 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N Synonym: chromium iii nitrate nonahydrate,unii-d2806iol1l,nitric acid, chromium 3+ salt, nonahydrate,acmc-20alg5,bis 7-chlorophenyl sulfone,cr.3no3.9h2o,ksc495a9j,chromium trinitrate, nonahydrate,chromium iii nitrate, nonahydrate,chromium 3+ nonahydrate trinitronate PubChem CID: 9887206 IUPAC Name: chromium(3+);trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Dual Chromium (+3, +6) Assurance™ Grade Speciation Standard, 30 mL in H2O, SPEX CertiPrep™
CAS: 7789-02-8 Molecular Formula: CrN3O9 Molecular Weight (g/mol): 238.01 InChI Key: PHFQLYPOURZARY-UHFFFAOYSA-N