Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Chromium Nitrate, Nonahydrate
SCI_ED

Catalog No. S25250A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25250A 500 g Reagent Grade
S25250 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25250A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7789-02-8
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Synonym chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Color Purple
Quantity 500 g
Linear Formula Cr(NO3)3·9H2O
Formula Weight 400.18
Grade Reagent
Packaging Reagent Grade
Density 1.8g/mL
Chemical Name or Material Chromium Nitrate, Nonahydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 5.1, III, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.