Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dual Chromium (+3, +6) Assurance™ Grade Speciation Standard, 30 mL in H2O, SPEX CertiPrep™
SDP

Catalog No. SPECDUALCR Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPECDUALCR 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPECDUALCR Supplier SPEX CertiPrep Supplier No. SPECDUALCR

missing translation for 'validMainframeMsg'

Includes

Certificate of Analysis with LC Chromatogram

Dual Chromium (+3, +6) Assurance™ Grade Speciation Standard for ICP and ICP-MS, 30 mL in H2O, Total Cr at 20 μg/mL, supplied with a complrehensive Certificate of Analysis including LC Chromatogra

  •  Dual Chromium (+3, +6) Speciation Standard
  • Total Cr at 20 μg/mL
  • 30 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Chemical Identifiers

CAS 7789-02-8 , 29595-25-3 , 7732-18-5
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N

Specifications

Description Dual Chromium (+3, +6) Speciation Standard
Packaging EA
CAS Min % 0.00%
CAS Max % 0.001%, 99.998%
Concentration Total Cr 20 µg/mL
Technique ICP-MS
Certifications/Compliance ISO/IEC 17025, ISO 17034, ISO 9001
UN Number 12352402
Quantity 1 Ea.
Chemical Name or Material Chromium (+3, +6)
ShelfLife 1 Year
Recommended Storage Ambient Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.