accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgII

CAS RN 7790-31-0

IUPAC Name: magnesium(2+) diiodide
Synonyms: magnesium iodide
Molecular Weight (g/mol): 278.11
Molecular Formula: I2Mg
InChi Key: BLQJIBCZHWBKSL-UHFFFAOYSA-L
SMILES: [Mg++].[I-].[I-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results