accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSClOO

CAS RN 7790-94-5

IUPAC Name: sulfurochloridic acid
Synonyms: chlorosulfonic acid chlorosulfuric acid
Molecular Weight (g/mol): 116.52
Molecular Formula: ClHO3S
InChi Key: XTHPWXDJESJLNJ-UHFFFAOYSA-N
SMILES: OS(Cl)(=O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)