Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorosulfonic Acid, Reagent, 95%, Spectrum™ Chemical
SDP

Catalog No. 18601630 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-630 500 mL Amber Glass Bottle
18-601-628 100 mL Amber Glass Bottle
18-601-629 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-630 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1231500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C1231, 7790-94-5, ClSO3H

Spectrum™ Chemical Chlorosulfonic Acid, Reagent , also known as Chlorosulfuric acid, is a powerful lachrymator and a synthetic organic reagent for the preparation of sulfonic acids. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7790-94-5
Molecular Formula ClHO3S
Molecular Weight (g/mol) 116.52
InChI Key XTHPWXDJESJLNJ-UHFFFAOYSA-N
IUPAC Name sulfurochloridic acid
SMILES OS(Cl)(=O)=O

Specifications

CAS Max % 100%
Quantity 500 mL
Packaging Amber Glass Bottle
UN Number UN1754
Percent Purity 95%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.