accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOHOHOHHOH2O

CAS RN 77938-63-7

IUPAC Name: 2,3,4,5,6-pentahydroxyhexanal hydrate
Synonyms: dextrose hydrate
Molecular Weight (g/mol): 198.17
Molecular Formula: C6H14O7
InChi Key: SPFMQWBKVUQXJV-UHFFFAOYNA-N
SMILES: O.OCC(O)C(O)C(O)C(O)C=O
Grade 
  • (1)
  • (3)
  • (1)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (5)
  • (3)
  • (4)
Percent Purity 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (9)
  • (4)
  • (9)
  • (1)
  • (3)
  • (1)
  • (8)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (4)
  • (5)