Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrose Monohydrate (Powder/USP), Fisher Chemical™

Catalog No. D15500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
3 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D15-500 500 g Poly Bottle
D15-3 3 kg Poly Pail
D15-12 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D15-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77938-63-7
Molecular Formula C6H14O7
Molecular Weight (g/mol) 198.17
MDL Number MFCD00149450
InChI Key SPFMQWBKVUQXJV-UHFFFAOYNA-N
Synonym d-glucose monohydrate, dextrose monohydrate, d-glucose, monohydrate, glucose monohydrate, d-+-glucose monohydrate, d +-glucose monohydrate, unii-lx22yl083g, dextrosum glucosum monohydricum, glucose water, dextrose hydrate
PubChem CID 22814120
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal hydrate
SMILES O.OCC(O)C(O)C(O)C(O)C=O

Specifications

Melting Point 83°C
Color White
Density 1.54g/mL
Packaging Poly Bottle
Linear Formula CH2OHCHO(CHOH)3CHOH·H2O
Quantity 500 g
Residual Solvents Meets Requirements (USP)
Vapor Pressure Negligible
Formula Weight 198.17
Grade USP
Physical Form Solid
Chemical Name or Material Dextrose Monohydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.