accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78-19-3

IUPAC Name: 3,9-diethenyl-2,4,8,10-tetraoxaspiro[5.5]undecane
Synonyms: pentaerythritol diacrolein acetal
Molecular Weight (g/mol): 212.25
Molecular Formula: C11H16O4
InChi Key: OOXMQACSWCZQLX-UHFFFAOYSA-N
SMILES: C=CC1OCC2(CO1)COC(OC2)C=C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)