Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane 98%, Thermo Scientific™

Catalog No. AC184670050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC184670050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC184670050 Supplier Thermo Scientific Chemicals Supplier No. 184670050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-19-3
Molecular Formula C11H16O4
Molecular Weight (g/mol) 212.25
MDL Number MFCD00006564
InChI Key OOXMQACSWCZQLX-UHFFFAOYSA-N
Synonym 3,9-divinyl-2,4,8,10-tetraoxaspiro 5.5 undecane, 3,9-divinylspirobi m-dioxane, diallylidene pentaerythritol, diallylidenepentaerythritol, pentaerythritol diacrolein acetal, acrolein pentaerythritol bisacetal, acrolein, cyclic diacetal with pentaerythritol, acrolein-pentaerythritol dicyclic acetal, acrolein, cyclic neopentanetetrayl acetal, 2,4,8,10-tetraoxaspiro 5.5 undecane, 3,9-diethenyl
PubChem CID 66218
IUPAC Name 3,9-diethenyl-2,4,8,10-tetraoxaspiro[5.5]undecane
SMILES C=CC1OCC2(CO1)COC(OC2)C=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 41°C to 45°C
Color White
Density 1.25
Infrared Spectrum Authentic
Quantity 5 g
Beilstein 19, IV, 5679
Formula Weight 212.24
Percent Purity ≥97.5%
Physical Form Crystalline Mass or Crystals
Chemical Name or Material 3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.