accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78-30-8

IUPAC Name: tris(2-methylphenyl) phosphate
Synonyms: tricresyl phosphate
Molecular Weight (g/mol): 368.37
Molecular Formula: C21H21O4P
InChi Key: YSMRWXYRXBRSND-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)