Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-o-tolyl Phosphate 98%, Thermo Scientific™

Catalog No. AC422295000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC422295000 500 g
AC422291000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422295000 Supplier Thermo Scientific Chemicals Supplier No. 422295000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-30-8
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.37
MDL Number MFCD00041906
InChI Key YSMRWXYRXBRSND-UHFFFAOYSA-N
Synonym tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
PubChem CID 6527
IUPAC Name tris(2-methylphenyl) phosphate
SMILES CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C

Specifications

Color Colorless to Yellow
Density 1.2
Infrared Spectrum Authentic
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Quantity 500 g
Merck Index 15, 9941
Formula Weight 368.36
Percent Purity ≥97.5%
Physical Form Viscous Liquid
Chemical Name or Material Tri-o-tolyl phosphate, 98%
Hazard Category GHS Signal Word: Danger
EINECSNumber 201-103-5
RTECSNumber TD0350000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.