accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3COH3COH3C

CAS RN 78-39-7

IUPAC Name: 1,1,1-triethoxyethane
Synonyms: triethyl orthoacetate
Molecular Weight (g/mol): 162.23
Molecular Formula: C8H18O3
InChi Key: NDQXKKFRNOPRDW-UHFFFAOYSA-N
SMILES: CCOC(C)(OCC)OCC
Molecular Weight (g/mol) 
  • (8)
  • (8)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (6)
  • (3)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
Boiling Point 
  • (5)
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (1)
  • (2)
  • (8)
  • (8)
  • (8)
  • (2)
  • (2)
  • (2)
  • (6)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (5)
  • (3)
  • (3)
  • (2)
  • (7)
  • (1)