Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl orthoacetate

Catalog No. T60402500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T60402500ML 500 mL
T60402100ML 100 mL
T604022L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T60402500ML Supplier Sigma Aldrich Supplier No. T60402500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triethyl orthoacetate can be used for the synthesis of:
  • Ordered mesoporous carbons for catalysis, electrochemistry and hydrogen storage applications
  • Esters of phosphonic acids, carboxylic acids and phosphinic acids
  • Ethyl alk-4-enoates by johnson-claisen rearrangement reaction
  • Benzoxazoles, benzimidazoles and oxazolo[4,5-b]pyridines
  • Quinazolin-4(3h)-one derivatives

Specifications

CAS 78-39-7
Density 0.885 g/mL (at 25°C (literature))
Boiling Point 142°C (lit.)
Molecular Formula C8H18O3
Refractive Index n20/D 1.396 (literature)
Linear Formula CH3C(OC2H5)3
MDL Number MFCD00009223
Quantity 500 mL
Synonym 1,1,1-Triethoxyethane
Molecular Weight (g/mol) 162.23
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.