CAS RN 78-78-4
2-Methylbutane, 99+%
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane, 99+%, extra pure
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane, 99+%, Extra Dry, AcroSeal™
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane 97.0+%, TCI America™
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane, for HPLC
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane, 99+%, for spectroscopy
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane, MilliporeSigma™
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane, Puriss p.a., ≥99.5% (GC), Solstice
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane, Reagent Grade, ≥99%, Solstice
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane Macron Fine Chemicals™
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
Avantor J.T.Baker 2-Methylbutane, BAKER™, J.T. Baker™
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N Synonym: isopentane,isoamylhydride,butane, 2-methyl,dimethylethylmethane,ethyldimethylmethane,1,1,2-trimethylethane,iso-pentane,1,1-dimethylpropane,butanes,iso-c5h12 PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
2-Methylbutane, SPEX CertiPrep™
CAS: 78-78-4 Molecular Formula: C5H12 Molecular Weight (g/mol): 72.15 MDL Number: MFCD00009338 InChI Key: QWTDNUCVQCZILF-UHFFFAOYSA-N PubChem CID: 6556 ChEBI: CHEBI:30362 IUPAC Name: 2-methylbutane SMILES: CCC(C)C
- 1
- 2