Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutane, Reagent Grade, ≥99%, Solstice

Catalog No. 60048074 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-074 500 mL Glass bottle
60-048-070 1 L Glass bottle
60-048-071 2.5 L Glass bottle
60-048-072 4 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-048-074 Supplier Solstice Supplier No. M32631500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Melting Point -160°C
Color Colorless
Density 0.62g/mL at 25°C
Boiling Point 28°C (1013 hPa)
Flash Point −51°C (−59.8°F)
Ignition Point 788°F
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)2
Quantity 500 mL
UN Number UN1265
Beilstein 1730723
Solubility Information Insoluble in water
Vapor Density 2.6 (vs air)
Vapor Pressure 11.17psi (20°C)
Formula Weight 72.15g/mol
Percent Purity ≥99%
Chemical Name or Material 2-Methylbutane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331
DOTInformation class 3 - PG 1 - Pentanes
RTECSNumber 201-142-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.