CAS RN 78-92-2
2-Butanol, anhydrous, 99%
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
sec-Butanol, 99%, Extra Pure
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
(+/-)-2-Butanol, 99%
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
sec-Butanol, 99+%, for analysis
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
2-Butanol, Solstice
CAS: 78-92-2 PubChem CID: 6568 ChEBI: CHEBI:35687
2-Butanol, anhydrous, 99%, packaged under Argon in resealable ChemSeal bottles
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
sec-Butanol, 99+%, Extra Dry, AcroSeal™
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
2-Butanol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
2-Butanol 99.0+%, TCI America™
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
2-Butanol, SPEX CertiPrep™
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
2-Butanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC), Solstice
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
Fisher Science Education™ Sec-Butyl Alcohol
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
2-Butanol, 98%, MilliporeSigma™
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O