Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanol, Solstice

Catalog No. 60090043 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
30 kg
Packaging:
Stainless Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-043 30 kg Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-043 Supplier Solstice Supplier No. 2412630KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sek.-Butylalcohol

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-92-2
PubChem CID 6568
ChEBI CHEBI:35687

Specifications

Quantity 30 kg
Flash Point 24 °C
Packaging Stainless Steel Can
Physical Form Liquid
Percent Purity min. 99 %
Grade Reagent
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.