accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN

CAS RN 78-97-7

IUPAC Name: 2-hydroxypropanenitrile
Synonyms: lactonitrile
Molecular Weight (g/mol): 71.08
Molecular Formula: C3H5NO
InChi Key: WOFDVDFSGLBFAC-UHFFFAOYNA-N
SMILES: CC(O)C#N
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results