accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CONONCH3OH3CNCH3H3CH3C(E/Z)

CAS RN 78416-81-6

IUPAC Name: hydrogen 9,10-dimethoxy-3-methyl-2-[(2,4,6-trimethylphenyl)imino]-2H,3H,4H,6H,7H-pyrimido[4,3-a]isoquinolin-4-one chloride
Synonyms: hydrogen trequinsin chloride
Molecular Weight (g/mol): 441.96
Molecular Formula: C24H28ClN3O3
InChi Key: DTCZZBVPTHVXFA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC2=C(C=C1OC)C1=CC(=NC3=C(C)C=C(C)C=C3C)N(C)C(=O)N1CC2
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results