accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONHClSOOONH2Na

CAS RN 78510-31-3

IUPAC Name: sodium 1-amino-4-[(4-chlorophenyl)amino]-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Synonyms: sodium 1-amino-4-[(4-chlorophenyl)amino]-9,10-dioxoanthracene-2-sulfonate
Molecular Weight (g/mol): 450.82
Molecular Formula: C20H12ClN2NaO5S
InChi Key: KINIBOSGQKLOIT-UHFFFAOYSA-M
SMILES: [Na+].NC1=C(C=C(NC2=CC=C(Cl)C=C2)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)