CAS RN 785708-33-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ T4
Modulator of alternative polyadenylation; inhibits rod photoreceptor gene expression via Nr2e3