accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78790-79-1

IUPAC Name: 3-aminopyridine-4-carbonitrile
Synonyms: 3-aminopyridine-4-carbonitrile
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: PEZNQSQPDQLHPN-UHFFFAOYSA-N
SMILES: NC1=C(C=CN=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results