CAS RN 79-34-5
1,1,2,2-Tetrachloroethane, 98.5%
CAS: 79-34-5 MDL Number: MFCD00000848 InChI Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N Synonym: s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan PubChem CID: 6591 ChEBI: CHEBI:36026 IUPAC Name: 1,1,2,2-tetrachloroethane SMILES: C(C(Cl)Cl)(Cl)Cl
1,1,2,2-Tetrachloroethane, MilliporeSigma™ Supelco™
Analytical standard suitable for chromatography and environmental analysis.
1,1,2,2-Tetrachloroethane, 98+%
CAS: 79-34-5 Molecular Formula: C2H2Cl4 Molecular Weight (g/mol): 167.838 MDL Number: MFCD00000848 InChI Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N Synonym: s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan PubChem CID: 6591 ChEBI: CHEBI:36026 IUPAC Name: 1,1,2,2-tetrachloroethane SMILES: C(C(Cl)Cl)(Cl)Cl
1,1,2,2-Tetrachloroethane, BAKER™, J.T. Baker™
CAS: 79-34-5 Molecular Formula: C2H2Cl4 Molecular Weight (g/mol): 167.838 InChI Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N Synonym: s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan PubChem CID: 6591 ChEBI: CHEBI:36026 IUPAC Name: 1,1,2,2-tetrachloroethane SMILES: C(C(Cl)Cl)(Cl)Cl
1,1,2,2-Tetrachloroethane, 98%, Spectrum™ Chemical
CAS: 79-34-5 Molecular Formula: C2H2Cl4 Molecular Weight (g/mol): 167.84 InChI Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N IUPAC Name: 1,1,2,2-tetrachloroethane SMILES: ClC(Cl)C(Cl)Cl
1,1,2,2-Tetrachloroethane, MilliporeSigma™
CAS: 79-34-5 Molecular Formula: C2H2Cl4 Molecular Weight (g/mol): 167.838 InChI Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N Synonym: s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan PubChem CID: 6591 ChEBI: CHEBI:36026 IUPAC Name: 1,1,2,2-tetrachloroethane SMILES: C(C(Cl)Cl)(Cl)Cl
1,1,2,2-Tetrachloroethane 97.0+%, TCI America™
CAS: 79-34-5 Molecular Formula: C2H2Cl4 Molecular Weight (g/mol): 167.838 MDL Number: MFCD00000848 InChI Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N Synonym: s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan PubChem CID: 6591 ChEBI: CHEBI:36026 IUPAC Name: 1,1,2,2-tetrachloroethane SMILES: C(C(Cl)Cl)(Cl)Cl
Methylene Blue Milk Smear Stain, Ricca Chemical
Modified Newman-Lampert Formulation, for Microscopic Bacterial Examination of Milk
1,1,2,2-Tetrachloroethane, (TCE), Reagents
CAS: 79-34-5 Molecular Formula: C2H2Cl4 Molecular Weight (g/mol): 167.84 InChI Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N Synonym: TCE, s-Tetrachloroethane, Acetylene tetrachloride IUPAC Name: 1,1,2,2-tetrachloroethane SMILES: ClC(Cl)C(Cl)Cl
Spex CertiPrep™ CLPV Series Volatile Analyte Mix A
Analytical standards for drinking water and raw source water
1,1,2,2-Tetrachloroethane, SPEX CertiPrep™
CAS: 79-34-5 Molecular Formula: C2H2Cl4 Molecular Weight (g/mol): 167.838 InChI Key: QPFMBZIOSGYJDE-UHFFFAOYSA-N PubChem CID: 6591 ChEBI: CHEBI:36026 IUPAC Name: 1,1,2,2-tetrachloroethane SMILES: C(C(Cl)Cl)(Cl)Cl