Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane, MilliporeSigma™

Catalog No. M8439481000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8439481000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M8439481000 Supplier MilliporeSigma Supplier No. 8.43948.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-34-5
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.838
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
Synonym s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
PubChem CID 6591
ChEBI CHEBI:36026
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Melting Point -42.5°C
Color Colorless
Density 1.59g/cm3 (25°C)
Boiling Point 146.35°C
Odor Suffocating
Packaging Glass Bottle
Quantity 1 L
UN Number UN 1702
Solubility Information Water: 2830g/L at 25°C (77°F)
Vapor Pressure 6.4 hPa (20°C)
Formula Weight 167.85
Percent Purity ≥99.0%
Grade Synthesis
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrachloroethane
Show More Show Less
Hazard Category H330: Fatal if inhaled.
Hazard Statement H310: Fatal in contact with skin.
Precautionary Statement H411: Toxic to aquatic life with long lasting effects.
EINECSNumber 201-197-8
RTECSNumber KI8575000
Recommended Storage 15° to 25°C (59° to 77°F); Tightly Closed. Dry. Keep in well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.