accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79-83-4

CH3H3CHOOHOHNOHO

CAS RN 79-83-4

IUPAC Name: 3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoic acid
Synonyms: pantothenic acid dihydrostreptomycin pantothenate
Molecular Weight (g/mol): 219.24
Molecular Formula: C9H17NO5
InChi Key: GHOKWGTUZJEAQD-UHFFFAOYNA-N
SMILES: CC(C)(CO)C(O)C(=O)NCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results