accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(R)POOOOO

CAS RN 79055-68-8

IUPAC Name: (2R)-2-azaniumyl-5-phosphonatopentanoate
Synonyms: (2R)-2-ammonio-5-phosphonatopentanoate
Molecular Weight (g/mol): 195.11
Molecular Formula: C5H10NO5P
InChi Key: VOROEQBFPPIACJ-SCSAIBSYSA-L
SMILES: [NH3+][C@H](CCCP([O-])([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)