Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ D-AP5
GSA_VAGREENER_CHOICESDP

Catalog No. 010650 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-065-0 50 mg
01-061 1 mg
01-061-0 10 mg
01-061-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 01-065-0 Supplier R&D Systems™ Supplier No. 0106/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective NMDA receptor antagonist; more active form of DL-AP5

Widely used competitive NMDA receptor antagonist. More active form of AP5. Also agonist at quisqualate-sensitized AP6 site where it is less potent than the L-isomer, L-AP5 (Cat. No.0107).

DL Mixture and L-isomer also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 79055-68-8
Molecular Formula C5H12NO5P

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 197.13
Purity >99%
Molecular Weight (g/mol) 197.127
Chemical Name or Material D-AP5
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.