accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COOONH3

CAS RN 79416-27-6

IUPAC Name: 5-methoxy-2,5-dioxopentan-1-aminium chloride
Synonyms: methyl aminolevulinate hcl methyl 5-aminolevulinate hcl methyl aminolevulinate hydrochloride
Molecular Weight (g/mol): 181.62
Molecular Formula: C6H12ClNO3
InChi Key: UJYSYPVQHFNBML-UHFFFAOYSA-N
SMILES: [Cl-].COC(=O)CCC(=O)C[NH3+]
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)