Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Aminolevulinate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A14111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14111G 1 g
A1411100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14111G Supplier TCI America Supplier No. A14111G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 79416-27-6 | C6H12ClNO3 | 181.616 g/mol

Chemical Identifiers

CAS 79416-27-6
Molecular Formula C6H12ClNO3
Molecular Weight (g/mol) 181.616
MDL Number MFCD00012868
InChI Key UJYSYPVQHFNBML-UHFFFAOYSA-N
Synonym methyl 5-amino-4-oxopentanoate hydrochloride, 5-aminolevulinic acid methyl ester hydrochloride, methyl aminolevulinate hydrochloride, methyl 5-aminolevulinate hydrochloride, metvixia, methyl aminolevulinate hcl, metvix pdt, 3-aminoacetylpropionic acid methyl ester hydrochloride, pentanoic acid, 5-amino-4-oxo-, methyl ester, hydrochloride, methyl aminolevulinate hydrochloride usan
PubChem CID 157921
ChEBI CHEBI:60641
IUPAC Name methyl 5-amino-4-oxopentanoate;hydrochloride
SMILES COC(=O)CCC(=O)CN.Cl

Specifications

Melting Point 122°C
Color White
Quantity 1 g
Formula Weight 181.62
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Aminolevulinate Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.