Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

δ-Aminolevulinic Acid Methyl Ester Hydrochloride, MP Biomedicals™

Catalog No. ICN15474891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15474891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15474891 Supplier MP Biomedicals, Inc Supplier No. 0215474891

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79416-27-6
Molecular Formula C6H12ClNO3
Molecular Weight (g/mol) 181.616
InChI Key UJYSYPVQHFNBML-UHFFFAOYSA-N
Synonym methyl 5-amino-4-oxopentanoate hydrochloride, 5-aminolevulinic acid methyl ester hydrochloride, methyl aminolevulinate hydrochloride, methyl 5-aminolevulinate hydrochloride, metvixia, methyl aminolevulinate hcl, metvix pdt, 3-aminoacetylpropionic acid methyl ester hydrochloride, pentanoic acid, 5-amino-4-oxo-, methyl ester, hydrochloride, methyl aminolevulinate hydrochloride usan
PubChem CID 157921
ChEBI CHEBI:60641
IUPAC Name methyl 5-amino-4-oxopentanoate;hydrochloride
SMILES COC(=O)CCC(=O)CN.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 181.62
Physical Form Crystalline Powder
Chemical Name or Material δ-Aminolevulinic Acid Methyl Ester Hydrochloride
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.