accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80-09-1

IUPAC Name: 4-(4-hydroxybenzenesulfonyl)phenol
Synonyms: bisphenol S diphone C 4,4'-bisphenol S bps 1 P,P'-dihydroxydiphenyl sulfone 4,4'-dihydroxydiphenyl sulfone
Molecular Weight (g/mol): 250.27
Molecular Formula: C12H10O4S
InChi Key: VPWNQTHUCYMVMZ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)